Learn More
Abcam CENTA, beta-lactamase substrate, Yellow chromogenic beta-lactamase substrate, 50MG
Catalog No. 502705097
Encompass_Preferred
MW 535.6 Da, Purity >98%. Yellow chromogenic β-lactamase substrate. Hydrolyses readily in the presence of all β-lactamases except for Aeromonas hydrophila metalloenzyme. Suitable for kinetic studies, the detection of the enzymes in crude extracts and chromatographic fractions. Absorbs light at 346 nm (Δε = -2500 M-¹ cm-¹) or 405 nm (Δε = +6400 M-¹ cm-¹).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5097 Supplier Abcam Supplier No. AB14562650MG
MW 535.6 Da, Purity >98%. Yellow chromogenic β-lactamase substrate. Hydrolyses readily in the presence of all β-lactamases except for Aeromonas hydrophila metalloenzyme. Suitable for kinetic studies, the detection of the enzymes in crude extracts and chromatographic fractions. Absorbs light at 346 nm (Δε = -2500 M-¹ cm-¹) or 405 nm (Δε = +6400 M-¹ cm-¹).
The product is subject to the following: Abcam Restricted Use Statement