Learn More
Aviva Systems Biology Corp Centa1 Antibody - C-terminal region (ARP52270_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52270_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Centa1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 4930431P11Rik, Centa1, GC1L, p42IP4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-285-680