Learn More
Labsciences Pipette Tips Standard 1-5ml, 500 count bag
Supplier:  Labsciences CEN6000
Encompass_Preferred
Blue/Non Sterile (Extra Long) 500/bag. Precision molded from virgin, chemically resistant and contamination free polypropylene. Sterile tips are sterilized by gamma irradiation in accordance with AMI guidelines.Tested for Fit and Function with most widely used pipettes on the market.
Catalog No. NC9743663