Learn More
Cayman Chemical Cnicin, Purity: ≥95%, CAS: 24394-09-0, 10 mg
SDP
Catalog No. 502917121
Encompass_Preferred
A germacranolide sesquiterpene lactone with diverse biological activities; irreversibly inhibits MurA (IC50s = 16.7 and 10.3 for the E. coli and P. aeruginosa enzymes, respectively) and is active against B. cereus and M. flavus (MICs = 1.25 and 5 µg/ml, respectively); inhibits the growth of MCF-7 and OVCAR-3 cancer cells (GI50s = 37.8 and 37.4 µM, respectively); reduces snake venom-induced paw edema in rats from 2.5-10 mg/kg
Catalog No. 50-291-7121 Supplier Cayman Chemical Supplier No. 3896910 MG
A germacranolide sesquiterpene lactone with diverse biological activities; irreversibly inhibits MurA (IC50s = 16.7 and 10.3 for the E. coli and P. aeruginosa enzymes, respectively) and is active against B. cereus and M. flavus (MICs = 1.25 and 5 µg/ml, respectively); inhibits the growth of MCF-7 and OVCAR-3 cancer cells (GI50s = 37.8 and 37.4 µM, respectively); reduces snake venom-induced paw edema in rats from 2.5-10 mg/kg