Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Checkers Center 3-Channel Rubber Duct Office Cable Protector

Catalog No. 19088441 Shop All Justrite Safety Group Products
Change view
Click to view available options
Length (English):
10 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-088-441 10 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-088-441 Supplier Checkers Supplier No. RFD310
Add to Cart
Add to Cart

Rubber duct systems protect and hide valuable cords and cables located in walkways or work areas. These non-metallic, flexible, industrial-grade rubber protectors are extremely durable, easy to install and organize portable cords and cables while reducing trip hazards.

  • Center Channel Sized To Fit Standard Power Cords And CAT5 Cable
  • Protector Must Be Slit For Standard Power Cords Due To Size Of Plug Connectors (Supplied Unslit)
  • Reduces Tripping Hazards
  • Protects And Organizes Portable Cords And Cables
  • Non-Metallic, Industrial-Grade Rubber Construction
  • Multiple Sizes For Various Cable Diameters
  • Grooved Underside For Better Traction
  • Optional Connector Kit Available For Most Sizes

Specifications

Length (English) 10 ft.
Length (Metric) 3 m
Color Black
Material Rubber
Product Type Cable Protector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.