Learn More
Grainger Center Drill Plain 1/32
Catalog No. 502765889
Encompass_Preferred
The Micro 100 Center Drill is a 1/32 inch diameter high speed steel center drill made in the USA. It has a straight shank and is designed for center drilling and spotting. The drill features a 118 degree split point tip for easy centering and smooth cutting. It can drill up to 1xD in steel and has an overall length of 2 inches. The center drill allows accurate starting of holes and is rigid for chatter-free operation. It's precision ground for concentricity within .0005 inches total indicator reading.
Catalog No. 50-276-5889 Supplier Grainger Supplier No. 16T466
The Micro 100 Center Drill is a 1/32 inch diameter high speed steel center drill made in the USA. It has a straight shank and is designed for center drilling and spotting. The drill features a 118 degree split point tip for easy centering and smooth cutting. It can drill up to 1xD in steel and has an overall length of 2 inches. The center drill allows accurate starting of holes and is rigid for chatter-free operation. It's precision ground for concentricity within .0005 inches total indicator reading.