Learn More
Grainger Center Drill Plain 1/32
Catalog No. 502767203
Encompass_Preferred
The Micro 100 Center Drill is a 1/32 inch diameter high speed steel center drill made in the USA. It has a straight shank and is designed for center drilling and spotting. The drill features a 118 degree split point tip for easy starting, smooth cutting, and accurate centering. It can be used on steel, cast iron, aluminum, brass, bronze, plastic and other materials. The drill has an overall length of 2 inches and comes with a bright finish for high visibility. It is ideal for precise centering and hole starting in milling, turning, grinding and other machining operations.
Catalog No. 50-276-7203 Supplier Grainger Supplier No. 16T485
The Micro 100 Center Drill is a 1/32 inch diameter high speed steel center drill made in the USA. It has a straight shank and is designed for center drilling and spotting. The drill features a 118 degree split point tip for easy starting, smooth cutting, and accurate centering. It can be used on steel, cast iron, aluminum, brass, bronze, plastic and other materials. The drill has an overall length of 2 inches and comes with a bright finish for high visibility. It is ideal for precise centering and hole starting in milling, turning, grinding and other machining operations.