Learn More
Grainger Center Drill Plain 1/32
Catalog No. 502768549
Encompass_Preferred
The Grainger center drill is a high-speed steel tool used to start holes for drilling on metals, plastics, and other materials. It has a 60 degree included angle point for starting holes accurately. The center drill comes in sizes ranging from 1/8 inch to 1/2 inch diameter. It has straight shank ends and is made in the USA. The center drill can be used in machining applications like milling and turning. It provides precise starting holes for drilling and produces clean, accurate holes. The high-speed steel construction allows faster speeds than carbon steel while retaining hardness.
Catalog No. 50-276-8549 Supplier Grainger Supplier No. 16T486
The Grainger center drill is a high-speed steel tool used to start holes for drilling on metals, plastics, and other materials. It has a 60 degree included angle point for starting holes accurately. The center drill comes in sizes ranging from 1/8 inch to 1/2 inch diameter. It has straight shank ends and is made in the USA. The center drill can be used in machining applications like milling and turning. It provides precise starting holes for drilling and produces clean, accurate holes. The high-speed steel construction allows faster speeds than carbon steel while retaining hardness.