Learn More
Grainger Center Drill Plain 1.60mm
Catalog No. 502768276
Encompass_Preferred
The Grainger center drill is a high-speed steel center drill with a 60 degree included angle point. It has a straight shank and is available in sizes ranging from 1/8 inch to 1 inch diameter. The center drills are made in the USA and have an overall length of 3 inches. They can be used for starting holes in preparation for drilling and provide an accurate starting point that prevents drill wandering. The center drills feature a polished finish for smooth cutting action.
Catalog No. 50-276-8276 Supplier Grainger Supplier No. 16T476
The Grainger center drill is a high-speed steel center drill with a 60 degree included angle point. It has a straight shank and is available in sizes ranging from 1/8 inch to 1 inch diameter. The center drills are made in the USA and have an overall length of 3 inches. They can be used for starting holes in preparation for drilling and provide an accurate starting point that prevents drill wandering. The center drills feature a polished finish for smooth cutting action.