Learn More
Grainger Center Drill Plain 3/16
Catalog No. 502765103
Encompass_Preferred
The Grainger Approved Center Drill is a high-speed steel center drill with a plain shank. It has a 118 degree point angle and is designed for center drilling and spotting. The drill has a 1/4 inch shank diameter and is 3 inches overall length. It is suitable for use on steel, cast iron, aluminum, brass, bronze, and plastic. The drill provides accurate starts and precise centering. Made in the USA, it meets ANSI standards for performance and durability.
Catalog No. 50-276-5103 Supplier Grainger Supplier No. 16T471
The Grainger Approved Center Drill is a high-speed steel center drill with a plain shank. It has a 118 degree point angle and is designed for center drilling and spotting. The drill has a 1/4 inch shank diameter and is 3 inches overall length. It is suitable for use on steel, cast iron, aluminum, brass, bronze, and plastic. The drill provides accurate starts and precise centering. Made in the USA, it meets ANSI standards for performance and durability.