Learn More
Grainger Center Drill Plain 3/64
Catalog No. 502768499
Encompass_Preferred
The Grainger Approved 3/64" Center Drill is a high-speed steel center drill with a plain shank. It has a 118 degree split point tip for accurate starting and clean holes. The drill diameter is 3/64" and the overall length is 2-1/2". The shank diameter is 1/8". This center drill is designed for use on mills, lathes, drill presses and CNC machines. It can drill through materials like alloy steel, cast iron, non-ferrous metals, plastics and wood. The plain shank allows use in chucks, collets or with an arbor. Made in the USA, this high quality center drill delivers accurate performance.
Catalog No. 50-276-8499 Supplier Grainger Supplier No. 16T467
The Grainger Approved 3/64" Center Drill is a high-speed steel center drill with a plain shank. It has a 118 degree split point tip for accurate starting and clean holes. The drill diameter is 3/64" and the overall length is 2-1/2". The shank diameter is 1/8". This center drill is designed for use on mills, lathes, drill presses and CNC machines. It can drill through materials like alloy steel, cast iron, non-ferrous metals, plastics and wood. The plain shank allows use in chucks, collets or with an arbor. Made in the USA, this high quality center drill delivers accurate performance.