Learn More
Grainger Center Drill Plain 5/64
Catalog No. 502767344
Encompass_Preferred
The Micro 100 Center Drill is a 5/64 inch diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 2 inches. It is designed for center drilling and starting holes in steel, cast iron, aluminum, brass, bronze, and plastics. The split point tip provides fast, accurate starts and reduced thrust. The center drill has an estimated life of 150 to 200 holes in 1018 steel.
Catalog No. 50-276-7344 Supplier Grainger Supplier No. 16T489
The Micro 100 Center Drill is a 5/64 inch diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 2 inches. It is designed for center drilling and starting holes in steel, cast iron, aluminum, brass, bronze, and plastics. The split point tip provides fast, accurate starts and reduced thrust. The center drill has an estimated life of 150 to 200 holes in 1018 steel.