Learn More
Grainger Center Drill Plain 5/64
Catalog No. 502767603
Encompass_Preferred
The Micro 100 Center Drill is a 5/64 inch diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 2 inches. It is designed for center drilling applications in steel, cast iron, aluminum, brass, bronze, and plastics. The center drill features a precision ground split point tip for accurate starting and minimum wander. It can be used in light duty machining applications. The center drill provides good chip control and tool life.
Catalog No. 50-276-7603 Supplier Grainger Supplier No. 16T468
The Micro 100 Center Drill is a 5/64 inch diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 2 inches. It is designed for center drilling applications in steel, cast iron, aluminum, brass, bronze, and plastics. The center drill features a precision ground split point tip for accurate starting and minimum wander. It can be used in light duty machining applications. The center drill provides good chip control and tool life.