Learn More
Grainger Center Drill Plain 5/64
Supplier:  Grainger 16T490
Encompass_Preferred
The Micro 100 Center Drill is a 5/64" diameter high speed steel center drill made in the USA. It has a straight shank and is designed for center drilling and spotting. The drill features a 118 degree split point tip for easy centering and smooth operation. It can drill up to 1xD in steel and has an overall length of 2-1/2". The center drill is precision ground for accuracy.
Catalog No. 50-276-5618