Learn More
Grainger Center Drill Plain 7/32
Catalog No. 502767401
Encompass_Preferred
The Micro 100 Center Drill is a 7/32" diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 3" and it is designed for center drilling applications in steel, cast iron, aluminum, brass, bronze, and plastics. This is a high quality center drill well-suited for precise drilling applications.
Catalog No. 50-276-7401 Supplier Grainger Supplier No. 16T498
The Micro 100 Center Drill is a 7/32" diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 3" and it is designed for center drilling applications in steel, cast iron, aluminum, brass, bronze, and plastics. This is a high quality center drill well-suited for precise drilling applications.