Learn More
Grainger Center Drill Plain 7/32
Catalog No. 502768495
Encompass_Preferred
The Micro 100 Center Drill is a 7/32" diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 2-1/2" and it is designed for center drilling applications in steel, cast iron, aluminum, brass, bronze and other materials. The center drill features a precision ground split point tip to provide fast, accurate starts with reduced thrust. It can be used in automatic screw machines, turret lathes, engine lathes, milling machines and machining centers.
Catalog No. 50-276-8495 Supplier Grainger Supplier No. 16T472
The Micro 100 Center Drill is a 7/32" diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 2-1/2" and it is designed for center drilling applications in steel, cast iron, aluminum, brass, bronze and other materials. The center drill features a precision ground split point tip to provide fast, accurate starts with reduced thrust. It can be used in automatic screw machines, turret lathes, engine lathes, milling machines and machining centers.