Learn More
Grainger Center Drill Plain 7/32
Catalog No. 502767342
Encompass_Preferred
The Micro 100 Center Drill is a 7/32" diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 2-1/2" and it is designed for center drilling applications in metalworking. The center drill features a precision ground split point tip to help prevent walking. It can be used on mills, lathes, drill presses, and CNC machines. The center drill is sold individually.
Catalog No. 50-276-7342 Supplier Grainger Supplier No. 16T497
The Micro 100 Center Drill is a 7/32" diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 2-1/2" and it is designed for center drilling applications in metalworking. The center drill features a precision ground split point tip to help prevent walking. It can be used on mills, lathes, drill presses, and CNC machines. The center drill is sold individually.