Learn More
Grainger Center Drill Plain 7/64
Catalog No. 502765893
Encompass_Preferred
The Micro 100 Center Drill is a 7/64" diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 2-1/2" and it is designed for center drilling applications in steel, cast iron, aluminum, brass, bronze and plastics. The center drill features a precision ground split point tip to provide fast, accurate starts with reduced thrust. It can be used in automatic screw machines, turret lathes, engine lathes, milling machines and drill presses.
Catalog No. 50-276-5893 Supplier Grainger Supplier No. 16T469
The Micro 100 Center Drill is a 7/64" diameter high speed steel center drill made in the USA. It has a straight shank, 118 degree split point tip, and is uncoated. The overall length is 2-1/2" and it is designed for center drilling applications in steel, cast iron, aluminum, brass, bronze and plastics. The center drill features a precision ground split point tip to provide fast, accurate starts with reduced thrust. It can be used in automatic screw machines, turret lathes, engine lathes, milling machines and drill presses.