Learn More
Grainger Keo Center Drill, Radius, 60°, 1.25 mm, High Speed Steel
Catalog No. 502772529
Encompass_Preferred
This radius type center drill is designed for precision machining applications requiring a 60° countersink angle. Constructed from high speed steel, this tool provides durability and reliable performance for right-hand cutting tasks.
  • Radius style countersink
  • 60° included angle
  • 1.25 mm drill point diameter
  • High speed steel construction
  • Bright uncoated finish
  • Two-flute spiral design
  • 3.15 mm overall length
Catalog No. 50-277-2529 Supplier Grainger Supplier No. 13X580
This radius type center drill is designed for precision machining applications requiring a 60° countersink angle. Constructed from high speed steel, this tool provides durability and reliable performance for right-hand cutting tasks.
  • Radius style countersink
  • 60° included angle
  • 1.25 mm drill point diameter
  • High speed steel construction
  • Bright uncoated finish
  • Two-flute spiral design
  • 3.15 mm overall length