Learn More
Grainger Center Drill Set Carbide Fractional Inch
Catalog No. 502767601
Encompass_Preferred
The Micro 100 Center Drill is a 1/8 inch diameter high speed steel center drill made in the USA. It has a straight shank and is designed for center drilling and spotting. The drill features a 118 degree split point tip for easy centering and smooth cutting. It can drill to a depth of 1-1/4 inches and has an overall length of 3-1/2 inches. The center drill is precision ground for accuracy and has an AlTiN nano coating to enhance tool life and lubricity. It works well in most materials including steel, cast iron, aluminum and more. This quality center drill is ideal for precise centering in milling and turning applications.
Catalog No. 50-276-7601 Supplier Grainger Supplier No. 16T464
The Micro 100 Center Drill is a 1/8 inch diameter high speed steel center drill made in the USA. It has a straight shank and is designed for center drilling and spotting. The drill features a 118 degree split point tip for easy centering and smooth cutting. It can drill to a depth of 1-1/4 inches and has an overall length of 3-1/2 inches. The center drill is precision ground for accuracy and has an AlTiN nano coating to enhance tool life and lubricity. It works well in most materials including steel, cast iron, aluminum and more. This quality center drill is ideal for precise centering in milling and turning applications.