Learn More
MSC Jergens Center Pull Hoist Ring, Alloy Steel, 10,000 lb Capacity, 1-8 Thread Size, 1 ea
Catalog No. 5000513892
Encompass
This safety engineered center pull hoist ring is designed for secure heavy-duty material lifting applications. Constructed from durable alloy steel with a black oxide finish, it is engineered for pulling operations and offers a maximum load capacity of 10,000 lbs. The component is inspected via magnetic particle methods and conforms to industry standards for reliability.
  • Maximum load capacity of 10,000 lbs
  • Constructed from high-strength alloy steel with a black oxide finish
  • Magnetic particle inspected for U-bars, bolts, pins, bases, washers, and bushings
  • Compatible with 1-8 thread size
  • Conforms to ASME B30.26 and MIL-STD-209K
  • Pivot and swivel functionality
Catalog No. 50-005-13892 Supplier MSC Supplier No. 72138118
This safety engineered center pull hoist ring is designed for secure heavy-duty material lifting applications. Constructed from durable alloy steel with a black oxide finish, it is engineered for pulling operations and offers a maximum load capacity of 10,000 lbs. The component is inspected via magnetic particle methods and conforms to industry standards for reliability.
  • Maximum load capacity of 10,000 lbs
  • Constructed from high-strength alloy steel with a black oxide finish
  • Magnetic particle inspected for U-bars, bolts, pins, bases, washers, and bushings
  • Compatible with 1-8 thread size
  • Conforms to ASME B30.26 and MIL-STD-209K
  • Pivot and swivel functionality