Learn More
Ampco Center Punch
SDP
Catalog No. 19174200 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Diameter (Metric):
8 mm
10 mm
13 mm
14 mm
16 mm
18 mm
20 mm
24 mm
27 mm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
19-174-200 27 mm
19-174-192 8 mm
19-174-193 10 mm
19-174-194 13 mm
19-174-195 14 mm
19-174-196 16 mm
19-174-197 18 mm
19-174-198 20 mm
19-174-199 24 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-174-200 Supplier Ampco Supplier No. P1297
Add to Cart
Add to Cart
Non-sparking tool designed to mark precise points on metal surfaces with lifetime warranty.
  • Bronze cylindrical center punch pin with a tapered end to center point.
  • Comes with lifetime warranty.
Specifications
Product Type Center Punch
Length (English) 9 -1/2 in.
Diameter (Metric) 27 mm
Quantity 1 Ea.