Learn More
MSC 80/20 Inc. Base Plate, 3-Hole, 15 Series, Aluminum, 1 ea.
Supplier: MSC 31445133
This center tap base plate is a milled aluminum component designed for use with 15 series profiles. The unit features an anodized finish for durability and includes a 3/8-16 center tap, making it suitable for secure mounting applications in framing systems.
- Made of 6063-T6 aluminum
- Milled process with an anodized finish
- Features three mounting holes for 5/16 in. socket head cap screws
- Compatible with 1515 and 1530 double profiles
- Adjustable mounting design
- ESD safe construction