Learn More
Thermo Scientific CENTER TUBE 1.5MM
Supplier:  Thermo Scientific 842312051951
Encompass
Centre Tube 1.5mm ID (EMT Torch); Centre tube intended for use with aqueous samples containing low - moderate levels of dissolved solids; Supplied as standard on all Radial iCAP instruments and can only be used with the centre tube holder for the EMT torch.
Catalog No. NC9071103