Learn More
Heidolph™ Centering Ring
SDP
Catalog No. 13900228 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Carousel 6 Plus Reaction System
Catalog No. For Use With (Equipment)
13-900-228 Carousel 6 Plus Reaction System
1 options
This item is not returnable. View return policy
Catalog No. 13-900-228 Supplier Heidolph™ Supplier No. 036210770
Add to Cart
Add to Cart
This item is not returnable. View return policy
Heidolph™ Centering Rings offer a leak-tight seal between glass reaction flasks and wide neck reflux tubes
  • Unique self-centering PTFE ring offers a leak-tight seal between the glass flask and wide neck reflux tube
  • For use with Carousel 6 Plus Reaction System glassware
Specifications
Material PTFE
Type Centering Ring
For Use With (Equipment) Carousel 6 Plus Reaction System
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov