Learn More
METTLER TOLEDO Centering Ring for Burette for EVA V1/V3 KF Titrator
Catalog No. 014388504 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-438-8504 2 mL
01-438-8521 5 mL
01-438-8538 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-438-8504 Supplier METTLER TOLEDO Supplier No. 30869326
Add to Cart
Add to Cart
Elevate your analysis with our specialized accessories for the EVA Karl Fischer V1 and V3 Titrators.
  • Accessories for the EVA Karl Fischer EVA V1 and V3 Titrators will simplify the handling of challenging samples, ensuring precise results.
  • Customize your Karl Fischer process for greater convenience and flexibility.
TRUSTED_SUSTAINABILITY
Specifications
Type Titration Burette
Volume (Metric) 2 mL