Learn More
Gardner Denver Welch Vacuum Tech Inc Centering ring NW16
SDP
Catalog No. NC1743035
Encompass_Preferred
This item has a minimum qty of 5 per supplier requirements.
Stainless steel centering ring with o-ring. Replaceable Fluoroelastomer 0-ring seal.
Catalog No. NC1743035 Supplier Gardner Denver Welch Vacuum Tech Inc Supplier No. 303101
This item has a minimum qty of 5 per supplier requirements.
Stainless steel centering ring with o-ring. Replaceable Fluoroelastomer 0-ring seal.