Learn More
MTC Bio 5mL Round Bottom Snap Cap Tube
Catalog No. NC2853776
Encompass_Preferred
Round bottom snap cap tubes adopt the conveniences of popular 1.5mL microcentrifuge tubes in higher capacities of 5mL, 7mL, and 10mL. Prevent a common source of cross-contamination in the lab by completely eliminating the threat of mismatched caps. These durable, polypropylene tubes are supplied sterile and are autoclavable. All tubes feature molded graduations and the 5 and 7mL tubes have frosted marking areas. Tubes are packed in easy-to-open sterile bags of 100.• Microtube convenience in larger capacities• Attached caps seal at the rim with a "click"• Sterile, easy-open bags of 100 tubes• Molded graduations and frosted marking area
Catalog No. NC2853776 Supplier MTC Bio Supplier No. C0905
Round bottom snap cap tubes adopt the conveniences of popular 1.5mL microcentrifuge tubes in higher capacities of 5mL, 7mL, and 10mL. Prevent a common source of cross-contamination in the lab by completely eliminating the threat of mismatched caps. These durable, polypropylene tubes are supplied sterile and are autoclavable. All tubes feature molded graduations and the 5 and 7mL tubes have frosted marking areas. Tubes are packed in easy-to-open sterile bags of 100.• Microtube convenience in larger capacities• Attached caps seal at the rim with a "click"• Sterile, easy-open bags of 100 tubes• Molded graduations and frosted marking area