Learn More
Chemglass Life Sciences Centrifuge Tube, 30mL, Free Standing, 29 x 80mm, Polypropylene, Sterile
SDP
Catalog No. 501979484
Encompass_Preferred
Centrifuge Tube, 30mL, Free Standing, 29 x 80mm, Polypropylene, Sterile. Conical centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86˚C. The maximum centrifugal force is 7,000xg. Polyethylene caps have a flat top surface for easy marking and labeling. CLS-4304-030 features a ‘skirted’ bottom that allow these tubes to stand upright without a separate test tube rack.
Catalog No. 50-197-9484 Supplier Chemglass Life Sciences Supplier No. CLS4304030
Centrifuge Tube, 30mL, Free Standing, 29 x 80mm, Polypropylene, Sterile. Conical centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86˚C. The maximum centrifugal force is 7,000xg. Polyethylene caps have a flat top surface for easy marking and labeling. CLS-4304-030 features a ‘skirted’ bottom that allow these tubes to stand upright without a separate test tube rack.