Learn More
Chemglass Life Sciences Centrifuge Tube, 50mL, 29 x 115mm, Polypropylene, Individually Wrapped, Sterile
SDP
Catalog No. 501979483
Encompass_Preferred
Centrifuge Tube, 50mL, 29 x 115mm, Polypropylene, Individually Wrapped, Sterile. Conical 50ml centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86˚C. The maximum centrifugal force is 12,000xg for the 15mL tubes and 15,000xg for the 50mL tubes*. Caps are made of polyethylene and have a flat top surface for easy marking or labeling.
Catalog No. 50-197-9483 Supplier Chemglass Life Sciences Supplier No. CLS4303W050
Centrifuge Tube, 50mL, 29 x 115mm, Polypropylene, Individually Wrapped, Sterile. Conical 50ml centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86˚C. The maximum centrifugal force is 12,000xg for the 15mL tubes and 15,000xg for the 50mL tubes*. Caps are made of polyethylene and have a flat top surface for easy marking or labeling.