Learn More
Chemglass Life Sciences Centrifuge Tube, 50mL, Self-Standing, Bagged, Sterile (25/Bag, 500/Case)
SDP
Catalog No. 501897796
Encompass_Preferred
Easy to see Black Graduation scaleLarge white writing areaEasy one-hand operationSterilized by E-Beam - DNase, RNase-free, non-pyrogenicRacked packaging is made using environmentally friendly re-usable racksHDPE material for the lid, PP material for the tubeRaw material complies with USP VI requirementsFlat markable cap with leak-proof sealMaximum centrifugation speed 12,000 x g
Catalog No. 50-189-7796 Supplier Chemglass Life Sciences Supplier No. CGN4304050
Easy to see Black Graduation scaleLarge white writing areaEasy one-hand operationSterilized by E-Beam - DNase, RNase-free, non-pyrogenicRacked packaging is made using environmentally friendly re-usable racksHDPE material for the lid, PP material for the tubeRaw material complies with USP VI requirementsFlat markable cap with leak-proof sealMaximum centrifugation speed 12,000 x g