Learn More
Beckman Coulter CENTFUGE TUBE/7/16X2 3/8/25PK
Catalog No. NC9806694
Encompass_Preferred
Thickwall Polyallomer Tube, 11 x 60mm, Volume: 3.0mL, Qty.: 25
Catalog No. NC9806694 Supplier Beckman Coulter Supplier No. 355636
Thickwall Polyallomer Tube, 11 x 60mm, Volume: 3.0mL, Qty.: 25