Learn More
Raven Environmental Products Centrifuge Tubes (19mm)-12
Catalog No. NC9171046
Encompass_Preferred
Raven conical bottom polycarbonate tube with percent graduations .02% to 100%. 19mm diameter. 11ml capacity. For use with Raven horizontal process centrifuge F-10300 or equivolent. Reusable. 12 per pack.
Catalog No. NC9171046 Supplier Raven Environmental Products Supplier No. B1010119
Raven conical bottom polycarbonate tube with percent graduations .02% to 100%. 19mm diameter. 11ml capacity. For use with Raven horizontal process centrifuge F-10300 or equivolent. Reusable. 12 per pack.