Learn More
Grainger Edwards Signaling Tone Generator, Zinc Alloy, 120 V AC, Indoor/Outdoor, 1 ea
Catalog No. 502789205
Encompass_Preferred
The 5540M Series central tone generator is a Division 2 rated device designed to produce a variety of signaling tones in indoor or outdoor environments. It is engineered for use in hazardous locations, including Class I, Division 2 and Class II, Division 2 areas, utilizing a specialized speaker or diaphragm to minimize the risk of sparking or arcing. This unit provides versatile signaling capabilities for industrial safety applications.
  • Features 55 selectable tone patterns
  • Compatible with multi-tone signals
  • Includes four inputs and four outputs
  • Designed with a zinc alloy housing
  • Rated for IP44 and NEMA 3R environments
  • Supports surface conduit mounting
  • Provides selectable volume levels
Catalog No. 50-278-9205 Supplier Grainger Supplier No. 16G665
The 5540M Series central tone generator is a Division 2 rated device designed to produce a variety of signaling tones in indoor or outdoor environments. It is engineered for use in hazardous locations, including Class I, Division 2 and Class II, Division 2 areas, utilizing a specialized speaker or diaphragm to minimize the risk of sparking or arcing. This unit provides versatile signaling capabilities for industrial safety applications.
  • Features 55 selectable tone patterns
  • Compatible with multi-tone signals
  • Includes four inputs and four outputs
  • Designed with a zinc alloy housing
  • Rated for IP44 and NEMA 3R environments
  • Supports surface conduit mounting
  • Provides selectable volume levels