Learn More
Beckman Coulter CENTRFUGE BOTTLS 16X76MM 25/PK
Catalog No. NC9197782
Encompass_Preferred
Bottle Assembly; Polycarbonate; 16 x 76mm; 10.4mL;
Catalog No. NC9197782 Supplier Beckman Coulter Supplier No. 355651
Bottle Assembly; Polycarbonate; 16 x 76mm; 10.4mL;