Learn More
Andwin Scientific CENTRIFUDGE TUBES 15ML
SDP
Catalog No. NC9794430
Encompass_Preferred
The 15 mL SuperClear™ Ultra-High Performance Centrifuge Tubes feature a thick-wall construction that permits high-speed operation. The tubes are also leakproof, providing a secure storage for samples. This product is free of RNase, DNase, and endotox
Catalog No. NC9794430 Supplier Andwin Scientific Supplier No. 21008216
The 15 mL SuperClear™ Ultra-High Performance Centrifuge Tubes feature a thick-wall construction that permits high-speed operation. The tubes are also leakproof, providing a secure storage for samples. This product is free of RNase, DNase, and endotox