Learn More
Electron Microscopy Sciences Centrifugal Filter Devices 2.0 ml, 0.22 µm, non-sterile
SDP
Supplier:  Electron Microscopy Sciences 6703169
Encompass
Spin-X centrifuge tube filters utilize centrifugation for the filtering of small volumes. The unit consists of a polypropylene filter insert and a centrifuge tube. It handles volumes up to 500ul. they have a cellulose acetate membrane for aqueous solutions which is ideal for the removal of bacteria, particles, and cells from media. They are also effective for DNA removal from agarose or acrylamide gels.
Spin-X LC units have a nylon membrane for applications such as HPLC sample preparation. Both have retailed volume of <5ul, and withstand relative centrifugal force (RFC) to 16,000xG.
Volume: 0.22 µm
Sterile: non-sterile
Catalog No. 50-309-23