Learn More
Med Vet International Centrifuge with 50ml x 6 Rotor
Catalog No. 503046166
Encompass_Preferred
Our newest addition to the Premiere low cost alternative centrifuge line is designed for the general lab. The sturdy design has large touch buttons for both speed (500 rpm increments) and timer (1 – 60 min.) settings. The bright and easy-to-read LCD display gives you the ability to see the speed and duration from further away. Capable of spinning specimens between 1,000 – 4,000 rpm, the sturdy base has a built-in safety feature that will not allow the lid to open while the rotor is in motion, and a beeping noise to indicate when spinning stops. Available in your choice of the two most popular rotor options to meet the needs of most common applications. Please order by model number based on the desired rotor.
Catalog No. 50-304-6166 Supplier Med Vet International Supplier No. XC2450
Our newest addition to the Premiere low cost alternative centrifuge line is designed for the general lab. The sturdy design has large touch buttons for both speed (500 rpm increments) and timer (1 – 60 min.) settings. The bright and easy-to-read LCD display gives you the ability to see the speed and duration from further away. Capable of spinning specimens between 1,000 – 4,000 rpm, the sturdy base has a built-in safety feature that will not allow the lid to open while the rotor is in motion, and a beeping noise to indicate when spinning stops. Available in your choice of the two most popular rotor options to meet the needs of most common applications. Please order by model number based on the desired rotor.