Learn More
Med Vet International Brushless Centrifuge 50ml x 6 Rotor with 15ml adapters
Catalog No. 503046060
Encompass_Preferred
With the versatile power of 2 centrifuges in one, the XC-2500 can spin 15ML and 50ML tubes — saving your customers diagnostic time, equipment cost, and counter-top space!FeaturesProgrammable settings with bright and easy-to-read LCD display.One touch panel for quick and easy timer settings.Fixed Angled Rotor with adapters for 6?×?15mL or 50mL tube holders.Built-in safety feature keeps lid closed while rotor is in use.Brushless motor for long lasting operation
Catalog No. 50-304-6060 Supplier Med Vet International Supplier No. XC2500
With the versatile power of 2 centrifuges in one, the XC-2500 can spin 15ML and 50ML tubes — saving your customers diagnostic time, equipment cost, and counter-top space!FeaturesProgrammable settings with bright and easy-to-read LCD display.One touch panel for quick and easy timer settings.Fixed Angled Rotor with adapters for 6?×?15mL or 50mL tube holders.Built-in safety feature keeps lid closed while rotor is in use.Brushless motor for long lasting operation