Learn More
USA Scientific Inc Centrifuge 5910 Ri touch interface refrigerated with Rotor S-4xUniversal incl universal buckets and adapters for 5 mL/15 mL/50 mL conical tubes and plates 120 V/50-60 Hz
Catalog No. 5000460760
Encompass
Centrifuge 5910 Ri touch interface refrigerated with Rotor S-4xUniversal incl universal buckets and adapters for 5 mL/15 mL/50 mL conical tubes and plates 120 V/50-60 Hz
Catalog No. 50-004-60760 Supplier USA Scientific Inc Supplier No. 40590343
Centrifuge 5910 Ri touch interface refrigerated with Rotor S-4xUniversal incl universal buckets and adapters for 5 mL/15 mL/50 mL conical tubes and plates 120 V/50-60 Hz