Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genesis BPS™ Centrifuge Balance

Catalog No. 22050479 Shop All Globe Scientific Products
Change view
Click to view available options
Length (Metric) Exterior:
32.02 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Exterior
22-050-479 32.02 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-050-479 Supplier Genesis BPS™ Supplier No. BPS428SE340
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Helps ensure matched centrifuge loads before processing

  • Helps reduce imbalance, vibration, and unnecessary stress on centrifuge equipment
  • Supports consistent blood component processing and laboratory workflow control
  • Simple bench-ready design makes balancing fast and repeatable for operators
  • Useful for blood banks, hospitals, and clinical laboratories handling centrifuged products
  • Helps protect equipment while supporting efficient daily processing routines

Specifications

Refrigerated No
Frequency 50/60 Hz
Voltage 100/115/220/230 V
Display Type LCD
Product Type Centrifuge Balance
Width (Metric) Exterior 18 cm
Length (Metric) Exterior 32.02 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.