Learn More
Bellco Glass, Inc. Centrifuge Bottle,590ml
SDP
Catalog No. 501945587
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Supplied with a 38-400 black phenolic rubber lined screw cap.. MADE IN THE USA Dimensions: 96x173mm Size: 590ml
Catalog No. 50-194-5587 Supplier Bellco Glass, Inc. Supplier No. 304500600
Manufactured from type 1 class a Borosilicate glass. Supplied with a 38-400 black phenolic rubber lined screw cap.. MADE IN THE USA Dimensions: 96x173mm Size: 590ml