Learn More
Bellco Glass, Inc. Centrifuge Bottle,780ml
SDP
Catalog No. 501945588
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Supplied with a 38-400 black phenolic rubber lined screw cap. Dimensions: 96x173mm Size: 780ml
Catalog No. 50-194-5588 Supplier Bellco Glass, Inc. Supplier No. 304500800
Manufactured from type 1 class a Borosilicate glass. Supplied with a 38-400 black phenolic rubber lined screw cap. Dimensions: 96x173mm Size: 780ml