Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter CENTRIFUGE BOTTLES (16 X 76 MM

Catalog No. NC9326754
Encompass

Tube, Thinwall; Polyallomer; 16 x 76mm; 13.5mL; 50/PK

Catalog No. NC9326754 Supplier Beckman Coulter Supplier No. 326814
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tube, Thinwall; Polyallomer; 16 x 76mm; 13.5mL; 50/PK

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.