Learn More
MTC Bio myFuge™ 5 Mini-Centrifuge, 1/EA
Catalog No. 501871035
Encompass
MyFuge™ 5 Mini-centrifuge for 4x5mL tubes, 5000rpm, 2000xg
Catalog No. 50-187-1035 Supplier MTC Bio Supplier No. C2595
MyFuge™ 5 Mini-centrifuge for 4x5mL tubes, 5000rpm, 2000xg