Learn More
Cole-Parmer Raven Environmental F-10300 Benchtop Centrifuge; 110 VAC 60 Hz
Catalog No. 50632418 Shop All Cole-Parmer Products
Encompass
Heavy-duty stainless steel and aluminum construction makes this benchtop centrifuge ideal for industrial environments such as food and beverage, wastewater plants, meat packing, and paper mills. Includes six polycarbonate reusable tubes clearly marked to indicate percent volume. The conical shape of the tubes accentuate small amounts of solids concentration to as low as 0.2%. Separate solids from liquid for improved laboratory tests.
Catalog No. 50-632-418 Supplier Cole-Parmer Supplier No. 0547710
Heavy-duty stainless steel and aluminum construction makes this benchtop centrifuge ideal for industrial environments such as food and beverage, wastewater plants, meat packing, and paper mills. Includes six polycarbonate reusable tubes clearly marked to indicate percent volume. The conical shape of the tubes accentuate small amounts of solids concentration to as low as 0.2%. Separate solids from liquid for improved laboratory tests.