Learn More
Med Vet International McKesson Microhematocrit Centrifuge 24 Place Horizontal Rotor Fixed Speed 12 000 RPM 603 Each
Catalog No. 503072338
Encompass_Preferred
McKesson Microhematocrit Centrifuge 24 Place Horizontal Rotor Fixed Speed 12 000 RPM 603 Each
Catalog No. 50-307-2338 Supplier Med Vet International Supplier No. 883760EA
McKesson Microhematocrit Centrifuge 24 Place Horizontal Rotor Fixed Speed 12 000 RPM 603 Each