Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS Centrifuge Multi 120 V FC5718R

Catalog No. 02112153 Shop All OHAUS Products
Change view
Click to view available options
Capacity:
4 x 200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
02-112-153 4 x 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-112-153 Supplier OHAUS Supplier No. 83042046
Earn Up to $2,000 in OHAUS Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Frontier FC5718R Refrigerated Centrifuge - Compact, High-Speed Performance for Every Lab Application

  • The Frontier FC5718R is a high-speed refrigerated centrifuge designed for multipurpose use across research, industrial, and clinical laboratory environments.
  • Maximum capacity of 4 x 200 mL and maximum relative centrifuge force of 23,542 x g.
  • Speed range of 200 to 18,000 rpm delivers outstanding centrifugation performance in a compact, bench-friendly footprint.
  • Part of the Frontier 5000 Multi-Pro series - a modular, scalable centrifugation platform designed to adapt to evolving laboratory needs.
  • Compatible with a wide variety of rotors and accessories to accommodate different tube types, sizes, and volumes.
  • The Frontier 5000 series spans 11 models with capacities from 0.2 mL to 4 x 1000 mL.
  • Supported applications include centrifugal separation, general sample preparation, cell pelleting, protein separation, nucleic acid separation, cell concentrating, chemical separation, diagnostics, environmental sample preparation, and blood sample processing.
  • User-friendly design features a backlit LCD display for clear visibility of run parameters.
  • Automatic rotor recognition detects the installed rotor and configures compatible operating parameters automatically.
  • Ten selectable acceleration and deceleration rates provide precise control for a wide range of applications.
  • Stores up to 99 programs for fast and efficient method recall.
  • Constructed with a corrosion-resistant stainless steel chamber for durability and protection against harsh reagents.
  • Polycarbonate/ABS plastic lid provides structural durability and visibility into the chamber.
  • Weighs 60 kg, offering stability while remaining lighter than comparable high-capacity models.
  • CFC-free refrigeration system cools from room temperature to 4°C in under 10 mins.
  • Operating temperature range from -20°C to +40°C (-4°F to +104°F) using environmentally responsible R290 refrigerant.
  • Continuous sample temperature indication enables monitoring during operation.
  • Rated for ambient operating conditions of 5°C to 35°C at up to 80% relative humidity, non-condensing.
  • Motor-driven lid-locking system ensures safe and secure operation.
  • Dual imbalance sensing protects both the instrument and samples from unbalanced loads.
  • Over-speed protection automatically enforces rotor speed limits.
  • Operates at a low noise level of ≤60 ±2 dB(A), suitable for noise-sensitive environments.
  • Power consumption is rated at 660 W, with AC power included.
  • Programmable run time from 10 sec. to 99 hr., 59 min., and 59 sec., with continuous operation capability.
  • Quick spin function enables rapid short-duration centrifugation.
  • CE and FCC certified for compliance with international safety and electromagnetic compatibility standards.
  • Dimensions: 731 mm (W) x 407 mm (D) x 359 mm (H) - 28.78 in. x 16.02 in. x 14.13 in.
  • Manufactured in Germany, delivering reliable high-speed refrigerated centrifugation for demanding laboratory workflows.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, FCC
Capacity 4 x 200 mL
Max. RCF 23,542 x g
Max. Speed 18,000 rpm
Refrigerated Yes
Green Features CFC-free refrigeration system, R290 refrigerant
Memory 99 Programs
Amperage 5.5 A
Product Type Centrifuge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.