Learn More
Unity Lab Services™ Centrifuge Plus Requalification Service with Cycle Testing
Shop All Thermo Scientific ProductsDescription
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
- Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
- Key tests Min/Max speed, Brief /Extended time, temperature for refrigerated models.
- Cycle testing (CT): Point-in-time test that monitors a unit and a specified rotor at predefined speed, time, and temperature (if applicable) set points.
- Testing demonstrates that a unit and specified rotor can meet customer specifications.
- Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
Specifications
Specifications
| For Use With (Equipment) | Centrifuge |
| Product Type | Requalification Service |