Learn More
Unity Lab Services™ Centrifuge Plus Requalification Service
Catalog No. 1673543 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Centrifuge
Catalog No. For Use With (Equipment)
16-735-43 Centrifuge
1 options
Catalog No. 16-735-43 Supplier Unity Lab Services™ Supplier No. RQCENPLUS
Add to Cart
Add to Cart
Requalification Services for Centrifuge Plus (documentation included)
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
  • Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
  • Key tests Min/Max speed, Brief /Extended time, temperature for refrigerated models.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Centrifuge
Product Type Requalification Service